Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Auris Enrichment Broth
SDP

Catalog No. 21200328 Shop All Hardy Diagnostics Products
This item is not returnable. View return policy
Catalog No. 21-200-328 Supplier Hardy Diagnostics™ Supplier No. R12
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enrichment broth recommended for the selective enrichment of C. auris from patient skin and environmental samples.

Auris Enrichment Broth is recommended for the selective enrichment of Candida auris from patient skin and environmental samples. The medium is based on the CDC formulation for Sabouraud Salt Dulcitol Broth with Chloramphenicol and Gentamicin.

Specifications

Form Custom Solutions
Manufacturing Origin United States
Sterility Non-sterile
Target Organism Class Multidrug-resistant, Gram-negative Organisms
Test Type Microbial Detection
Fill Volume 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.