Learn More
AdipoGen Aurofusarin
SDP
Supplier:  AdipoGen BVT0392M001
Encompass_Preferred
Chemical. CAS 13191-64-5. Formula C30H18O12. MW 570.5. Isolated from Fusarium graminearum. Dimeric naphthoquinone. Antibiotic activity. Mycotoxin. Toxic against mammalians and avian species. Analytical standard in food industry.
Catalog No. 50-256-1955
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.