Learn More
Proteintech Group Inc AuthentiKine™ Human IFN-beta ELISA Kit
SDP
Supplier:  Proteintech Group Inc KE0019596T
Encompass_Preferred
KE00195 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The IFN-beta ELISA kit is to be used to detect and quantify protein levels of endogenous IFN-beta. The assay recognizes human IFN-beta. An antibody specific for IFN-beta has been pre-coated onto the microwells. The IFN-beta protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody specific for IFN-beta is added to detect the captured IFN-beta protein. For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Catalog No. 50-225-8423