Learn More
Market Lab Auto Digital Sphygmomanometer
Supplier: Market Lab 3928
This clinically accurate sphygmomanometer meets the A/A rating according to the British Hypertension Society. It offers an easy-to-read digital display that includes pulse rate with an irregular heartbeat recognition. Automatically stores the last 99 readings, heart rate, and date/time. Runs on two AA-batteries (included) or an optional six-volt adapter.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.