Learn More
Bellco Glass, Inc. Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045
Supplier: Bellco Glass, Inc. 791035300
Manufactured from medium wall Borosilicate Glass Type 1 Class A, with permanent graduations designed for DNA/RNA protocols. Includes high temperature red PBT cap with PTFE faced silicone liner. Replacement cap 5639-90045