Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
GREENER_CHOICE

Catalog No. 21402360
Change view
Click to view available options
Volume (Metric):
20 μL
40 μL
50 μL
130 μL
250 μL
Filtered:
Filtered
Non-filtered
Tip Style:
Low Retention
Standard
Wide Bore
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Tip Style Fit for Purpose
21-402-360 250 μL Non-filtered Wide Bore Sterile
21-402-650 40 μL Filtered Standard Sterile and PCR Ready
21-402-651 50 μL Non-filtered Standard Sterile
21-402-652 50 μL Non-filtered Standard Certified Quality
21-402-190 20 μL Filtered Standard Sterile and PCR Ready
21-402-638 20 μL Filtered Low Retention Sterile and PCR Ready
21-402-191 20 μL Non-filtered Standard Sterile
21-402-636 20 μL Non-filtered Low Retention Sterile
21-402-192 20 μL Non-filtered Standard Certified Quality
21-402-193 130 μL Filtered Standard Sterile and PCR Ready
21-402-641 130 μL Filtered Low Retention Sterile and PCR Ready
14-754-324 130 μL Filtered Wide Bore Sterile and PCR Ready
21-402-194 250 μL Non-filtered Standard Sterile
21-402-639 250 μL Non-filtered Low Retention Sterile
21-402-195 250 μL Non-filtered Standard Certified Quality
21-402-640 250 μL Non-filtered Low Retention Certified Quality
14-754-325 250 μL Non-filtered Wide Bore Certified Quality
21-402-637 20 μL Non-filtered Low Retention Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 21-402-360 Supplier Thermo Scientific™ Supplier No. 919261G
Add to Cart
Add to Cart

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Wide Bore
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 4800 tips
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.