Learn More
Axygen™ Automation Tips for PerkinElmer™, 384-Well
GREENER_CHOICE
Catalog No. 14222367 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Filtered
14-222-367 Non-sterile Non-filtered
10-320-174 Sterile Filtered
14-222-368 Sterile Non-filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-222-367 Supplier Axygen™ Supplier No. PK384R
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 384-Well are racks of non-filtered, 30μL robotic pipette tips. Constructed of clear polypropyle to stringent specifications. Available in sterile and non-sterile options.
  • Tip format: 96 tip and 384 tip
  • Tip volume range: 20 μL to 235 μL
  • Tip material: Clear or black polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: MDT-96 P50, MDT-384-30 μL
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Order Info
Rack: 384 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Compliance
Axygen products are manufactured in a clean, controlled state of the art facility and certified RNase-, DNase- and endotoxin-safe
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Green
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) PerkinElmer Janus, PerkinElmer Evolution P3, PerkinElmer MiniTrak, PerkinElmer PlateTrak, PerkinElmer MDT-96 P235, PerkinElmer MDT-96 P50, PerkinElmer MDT-384 30μL
Material Polypropylene