Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
GREENER_CHOICE

Catalog No. 2140325
Change view
Click to view available options
Volume (Metric):
20 μL
30 μL
40 μL
50 μL
235 μL
Autoclavable:
Autoclavable
Not Autoclavable
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Autoclavable Filtered Fit for Purpose
21-403-25 235 μL Not Autoclavable Non-filtered Sterile
21-402-632 40 μL Not Autoclavable Filtered Sterile and PCR Ready
21-402-631 50 μL Autoclavable Non-filtered Certified Quality
21-403-26 235 μL Autoclavable Non-filtered Certified Quality
02-707-529 20 μL Not Autoclavable Filtered Sterile and PCR Ready
21-931-261 30 μL Not Autoclavable Non-filtered Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 21-403-25 Supplier Thermo Scientific™ Supplier No. 929261
Add to Cart
Add to Cart
This item is not returnable. View return policy

Increase pipetting effectiveness with these automation tips, achieving accurate and precise performance due to superior tip straightness and a low coefficient of variation.

Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

Features:

  • Available as sterile and filtered
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 235 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 4800 tips
For Use With (Equipment) Packard Liquid Handling System, PerkinElmer
Product Line PerkinElmer/Packard
Fit for Purpose Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.