Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Qiagen™ Workstations
GREENER_CHOICE

Catalog No. 21402229
Change view
Click to view available options
Volume (Metric):
250 μL
300 μL
1000 μL
1100 μL
Filtered:
Filtered
Non-filtered
Packaging:
96 Tips/Tray, 10 Trays/Case
96 Tips/Tray, 50 Trays/Case
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Packaging Fit for Purpose
21-402-229 1100 μL Non-filtered 96 Tips/Tray, 50 Trays/Case Sterile
14-754-338 250 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile and PCR Ready
21-402-232 300 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Sterile
21-402-233 300 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Certified Quality
02-707-537 1000 μL Filtered 96 Tips/Tray, 10 Trays/Case Sterile and PCR Ready
21-402-235 1100 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Sterile
21-402-236 1100 μL Non-filtered 96 Tips/Tray, 10 Trays/Case Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 21-402-229 Supplier Thermo Scientific™ Supplier No. 953253
Add to Cart
Add to Cart

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.

  • Available in nonsterile, sterile and with the ART™ filter technology
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Conductive
Packaging 96 Tips/Tray, 50 Trays/Case
For Use With (Equipment) Universal Fit, Qiagen
Product Line Qiagen
Fit for Purpose Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.