Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
GREENER_CHOICE

Catalog No. 02682273
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
150 μL
200 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Filtered Fit for Purpose
02-682-273 100 μL Non-sterile Non-filtered Certified Quality
21-403-428 50 μL Sterile Filtered Sterile and PCR Ready
21-403-434 150 μL Sterile Filtered Sterile and PCR Ready
21-403-432 200 μL Sterile Non-filtered Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-682-273 Supplier Thermo Scientific™ Supplier No. 907262
Add to Cart
Add to Cart

Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.

  • Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
  • Available in filtered, low retention or extended length for increased volume to reach into deep well plates
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Low Retention, Extended Length
For Use With (Equipment) Tecan Aquarius, Tecan TeMo
Product Line Tecan
Fit for Purpose Certified Quality
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.