Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Autophagy Antibody Pack
SDP

Catalog No. NB91094877 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB91094877 5 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB91094877 Supplier Novus Biologicals™ Supplier No. NB91094877

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Includes

  • 1.NB100-2220: LC3B/MAP1LC3B Antibody
  • 2.NB110-53818: ATG5 Antibody
  • 3.NB110-56893: ATG9A Antibody
  • 4.NB110-82384: ATG16L1 Antibody
  • 5.NB110-87318: Beclin 1/ATG6 Antibody

For Research applications

Autophagy specifically detects Autophagy in Alligator, Bovine, Human, Mouse, Pig, Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot.

Specifications

Antigen Autophagy
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot
Concentration 25ul vials of each of following: NB100-2220, NB110-87318, NB110-53818, NB110-56893 and NB110-82384.
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation See individual datasheets
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Gene Symbols BECN1
Immunogen See individual datasheets
Quantity 5 Vials
Research Discipline Autophagy
Gene ID (Entrez) 8678
Test Specificity Autophagy Antibodies (Beclin, LC3, ATG5, ATG9, ATG16L1)
Includes 1.NB100-2220: LC3B/MAP1LC3B Antibody
2.NB110-53818: ATG5 Antibody
3.NB110-56893: ATG9A Antibody
4.NB110-82384: ATG16L1 Antibody
5.NB110-87318: Beclin 1/ATG6 Antibody
Target Species Alligator, Bovine, Human, Mouse, Pig, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.