Description
- Anodized Aluminum tray with 1 calibration standard holder, 3 system suitability holders and 64 sample positions (standard 40 mL glass TOC vials)
- Automated XYZ arm with septum piercing probe, rinse station
- 100 to 240 VAC, 47 to 63 Hz
Specifications
Specifications
| Type | Autosampler |
| For Use With (Application) | Pharmaceuticals |
| Voltage | 100/240 VAC |