Learn More
Cytiva AutoSeq G-50 Columns
Catalog No. 45001401 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
250 Columns
50 Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-401 250 Columns
45-001-400 50 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-001-401 Supplier Cytiva Supplier No. 27534002
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Use for rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing. Cytiva Life Sciences™ illustra™ AutoSeq G-50 Columns provide effective purification essential for high-quality sequencing results.
  • Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
  • Columns that are ready to use enable sample application to collection of purified product in less than 4 min.
  • Can be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems
  • Typical sample volume 12 to 25 μl
  • Suitable for purification of DNA greater than 20 bases in length
  • Optimized for automated fluorescent sequencing
TRUSTED_SUSTAINABILITY
Specifications
Format Column
For Use With (Application) Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing
Product Type Column
Quantity 250 Columns