Learn More
Beckman Coulter Junior BCR-ABL Mutation Analysis Kit, CE
Supplier:  Beckman Coulter C15126
Encompass
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin-downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 x g. The rotor accommodates 18 x 1.5/2.0 mL microcentrifuge tubes. It has a compact footprint, digital display, 10 acceleration/deceleration rates, and a pulse key for short spin cycles. Safety features include an imbalance detector that prevents the rotor from spinning if improperly loaded.
Catalog No. NC2306603
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.