Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AUTS2 Monoclonal Antibody (CL7067)
GREENER_CHOICE

Catalog No. PIMA531446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531446 Supplier Invitrogen™ Supplier No. MA531446
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: GISPLPGGER FPYPSFHWD Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 89%.

AUTS2 also named as FBRSL2 or autism susceptibility candidate 2 is a 1259 amino acid protein, which belongs to the AUTS2 family. AUTS2 is highly expressed in brain, kidney and skeletal muscle. AUTS2 exists as four isoforms produced by alternative splicing events and is encoded by a gene located on human chromosome 7. The molecular weight of AUTS2 isoform is 65-70 kDa in the brain. The functional role of AUTS2 is not well known, although some studies have identified a putative role in transcriptional regulation during neuronal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AUTS2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL7067
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Auts2
Gene Accession No. A0A087WPF7
Gene Alias 2700063G02Rik; A730011F23Rik; autism susceptibility candidate 2; autism susceptibility gene 2 protein; Autism susceptibility gene 2 protein homolog; autism-related protein 1; AUTS2; D830032G16Rik; FBRSL2; Kiaa0442; MRD26
Gene Symbols Auts2
Host Species Mouse
Immunogen Synthetic peptide corresponding to Human AUTS2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319974
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.