Learn More
Heidolph™ AVA Software
SDP
Heidolph™ AVA Software easily automates chemistry experiments and procedures, includes one year of AVA Care Support
Supplier:  Heidolph™ 015200003
 View more versions of this product
Catalog No. 13-876-716
Only null left
Add to Cart
Request a Quote
Description
Description
  • AVA easily automates chemistry procedures
  • Makes chemistry safer by running unattended experiments safely, reducing manual errors, setting apparatus safety limits to prevent accidents
  • Configures audio and visual alarms
  • Advantages of software automation include improved productivity, and easy results sharing between users which improves research/collaboration
  • AVA software can:
    • Control, log common laboratory apparatus and processes
    • Eliminate time-consuming, manual data recording
    • Track what you do, as you do it and repeat in the experiment scheduler
    • Visualize results for rapid analysis and reporting
    • Link devices for dual control (pump and balance for additions)
    • Control up to four experiments in parallel
Specifications
Specifications
Level 3
1 Year of AVA Care Support
AVA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.