Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Avagacestat
GREENER_CHOICE

Catalog No. p-7161599
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-632-5 25 mg
63-635 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-632-5 Supplier Tocris Bioscience™ Supplier No. 6363/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent γ-secretase inhibitor; orally bioavailable

Avagacestat is a highly potent γ-secretase inhibitor (Aβ40 IC50 = 0.3 nM). Exhibits >137-fold selectivity for amyloid-β precursor protein (APP) processing over Notch. Reduces plasma and brain Aβ40 and Aβ42 levels in vivo. Orally bioavailable.

Specifications

Chemical Name or Material (2R)-2-[[(4-Chlorophenyl)sulfonyl][[2-fluoro-4-(1,2,4-oxadiazol-3-yl)phenyl]methyl]amino]-5,5,5-trifluoropentanamide
CAS 1146699-66-2
Synonym BMS-708163
Quantity 25 mg
Molecular Formula C20H17ClF4N4O4S
Formula Weight 520.89
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.