Learn More
Selleck Chemical LLC Avagacestat (BMS-708163) 5mg 1146699-66-2
Supplier: Selleck Chemical LLC S12625MG
Avagacestat (BMS-708163) is a potent, selective, orally bioavailable ?-secretase inhibitor of A?40 and A?42 with IC50 of 0.3 nM and 0.27 nM, demonstrating a 193-fold selectivity against Notch. Phase 2. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.