Learn More
Apexbio Technology LLC Avermectin B1a 65195-55-3 5mg
SDP
Supplier:  Apexbio Technology LLC A32035
Encompass_Preferred
Avermectin B1a (65195-55-3) is a macrocyclic lactone compound targeting -aminobutyric acid (GABA)-gated chloride channels It is designed to modulate these channels thereby inducing neuronal hyperpolarization and neuromuscular paralysis Avermectin B1a exerts its biological activity primarily through promoting chloride ion influx and inhibiting synaptic transmission in nematodes Experimental evidence shows that Avermectin B1a enhances binding of benzodiazepines (e g flunitrazepam) at GABA receptors with an EC50 approximately 50-fold lower compared to GABA Based on these pharmacological properties Avermectin B1a holds research potential in studies on anthelmintic and insecticidal mechanisms
Catalog No. 50-000-94535
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.