Learn More
Perkin Elmer US LLC AVI Calibration Methane Cell
Supplier:  Perkin Elmer US LLC L1250462
Encompass_Preferred
Discrepancies can exist between FT-IR spectrometers. An instrument's wavenumber and lineshape values can be adversely affected as a result of changing sampling accessories and components. Absolute Virtual Instrument (AVI) is a Spectrum software function that ensures wavelength and lineshape standardization for all FT-IR istruments. When AVI is turned on, the spectrometer's Spectrum software compares the spectrometer's wavelength and lineshape measurements against methane cell's measurements and updates the instrument's AVI profile, when necessary.
Catalog No. 50-211-2717
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.