Learn More
Avian Influenza A H4N6 Hemagglutinin Mouse anti-Virus, HRP, Antibody Pair, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Avian Influenza A H4N6 Hemagglutinin |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Gene Alias | Harvey rat sarcoma viral oncogene homolog |
| Host Species | Mouse |
| Quantity | 15 Plates |
| Regulatory Status | RUO |
| Target Species | Virus |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.