Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Avian Influenza A H7N9 Hemagglutinin Antibody Pair [HRP]
SDP

Catalog No. NB009883 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009883 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009883 Supplier Novus Biologicals™ Supplier No. NBP27942215PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Virus samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin.
Target Species Virus
For Use With (Application) Sandwich ELISA
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 110 ng of recombinant Avian Influenza A H7N9 Hemagglutinin.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-Avian Influenza A H7N9 Hemagglutinin monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL rabbit anti-Avian Influenza A H7N9 Hemagglutinin polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Avian Influenza A H7N9 Hemagglutinin
    Technique 1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.