Learn More
Description
Includes
- 1.NB100-1933: Avian Flu Matrix Protein 1 Antibody
- 2.NB100-56572: Avian Flu Nucleoprotein Antibody
- 3.NB100-56607: Avian Flu HA protein Antibody
- 4.NB100-56608: Avian Flu HA protein Antibody
- 5.NB100-56610: Avian Flu Matrix Protein 1 Antibody
- 6.NB100-56611: Avian Flu Nucleoprotein Antibody
Specifications
Specifications
| Antigen | Avian Influenza A Hemagglutinin |
| Applications | Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot |
| Gene Alias | ha; HA1; HA2; Hemagglutinin; Hemagglutinin ha1 chain; Hemagglutinin ha2 chain; Hemagglutinin precursor |
| Immunogen | Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 72-88 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 128-143 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 9-28 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 212-225 Format: Purified Rabbit IgG polyclonal antibody; Application: western blot; Immunogen: amino acids 58-77 Format: Purified Rabbit IgG polyclonal antibody; Application: ELISA, IF/ICC, IHC; Immunogen: amino acids 428-441 |
| Quantity | 6 Vials |
| Includes | 1.NB100-1933: Avian Flu Matrix Protein 1 Antibody 2.NB100-56572: Avian Flu Nucleoprotein Antibody 3.NB100-56607: Avian Flu HA protein Antibody 4.NB100-56608: Avian Flu HA protein Antibody 5.NB100-56610: Avian Flu Matrix Protein 1 Antibody 6.NB100-56611: Avian Flu Nucleoprotein Antibody |
| Target Species | Virus |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
