Learn More
Invitrogen™ Avian Influenza HA Type 4 Synthetic Peptide
Description
Avian Influenza Hemagglutinin 4 Synthetic Peptide, PEP-0412, from Invitrogen.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | Avian Influenza HA Type 4 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Lyophilized | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.