Learn More
Selleck Chemical LLC Fondaparinux Sodium (Org 31540) S3736-25mg
SDP
Supplier:  Selleck Chemical LLC S373625MG
Encompass_Preferred
Fondaparinux Sodium (Org 31540 Natural heparin pentasaccharide Fondaparin SR-90107A) is a synthetic glucopyranoside with antithrombotic activity It selectively binds to antithrombin III thereby potentiating the innate neutralization of activated factor X (Factor Xa) by antithrombin a synthetic inhibitor of Factor Xa
Catalog No. 50-311-0552
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.