Learn More
Miles Scientific AVICEL 500UM 20X20CM (25 PLATES/BOX)
SDP
Supplier:  Miles Scientific P05012
Encompass_Preferred
These plates are coated with AVICEL microcrystalline cellulose manufactured by the American Viscose division of FMC Corp. The adsorbent consists of regenerated alpha cellulose particles of nearly spherical shape and 50 micron mean diameter. Compounds separated on this cellulose tend to form more compact spots than on fibrous cellulose layers. Preparative TLC plates have adsorbent layers thicker than the 250 micron layers normally used for analytical separations. Thus, preparative plates can be used for separation of larger quantities of materials.The loading capacity for a given degree of sample component resolution increases roughly as the square root of the increased adsorbent layer thickness.
Catalog No. NC1027879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.