Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avidin Polyclonal Antibody, Rockland™
SDP

Catalog No. p-7190807
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1065 100 μg
50-199-1066 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1065 Supplier Rockland Immunochemicals Supplier No. 20041970100
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-AVIDIN is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Avidin [Hen Egg White]. No cross reactivity occurs against Streptavidin.

Avidin from chicken egg whites is a glycoprotein that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods. This protein contains four identical subunits having a combined molecular mass of 67,000 to 68,000 daltons. Each subunit binds one molecule of biotin, and studies have shown that tryptophan and lysine are involved in the binding site for biotin. The sequencing of the subunit indicates it consists of 128 amino acids. Avidin has an isoelectric point of 10-10.5 and is very soluble in water and salt solutions. Avidin is stable over a wide range of pH values and temperatures. Extensive chemical modification has little effect on the activity of avidin, making it useful for detection and protein purification. Pure avidin is a key component in certain immunohistochemistry (IHC) methods. Alkaline phosphatase (AP) and horseradish peroxidase (HRP) conjugates of avidin are useful for Western blotting or ELISA detection with appropriate enzyme substrates.

Specifications

Antigen Avidin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene AVD
Gene Accession No. P02701
Gene Alias AVD; avID; Avidin; mature avidin
Gene Symbols AVD
Host Species Rabbit
Immunogen Avidin (Hen Egg White)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396260
Target Species Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.