Learn More
MyBioSource Monoclonal IL-21 Antibody
SDP
Supplier:  MyBioSource MBS1563927
Encompass_Preferred
The MyBioSource monoclonal IL-21 antibody is a mouse IgG1 antibody raised against recombinant human IL-21. It is recommended for detection of IL-21 of human origin by ELISA, Western Blot, Immunohistochemistry, and Flow Cytometry. The antibody has been tested in ELISA and Western Blot applications. The immunogen is recombinant human IL-21 produced in E. coli. This monoclonal antibody is purified by affinity chromatography and has unknown cross-reactivity with other species. The antibody is provided at 100ug in PBS with 0.02% sodium azide and 0.5% BSA.
Catalog No. NC2480348
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.