Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVPR1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PI720364 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720364 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720364 Supplier Invitrogen™ Supplier No. 720364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Rabbit and Cat.

Vasopressin receptor 1a (AVPR1A) is a receptor for arginine vasopressin. The activity of this receptor is mediated by G-proteins which activate a phosphatidyl-inositol-calcium second messenger system. AVPR1A has been involved in social behaviors, including affiliation and attachment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AVPR1A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2
Gene AVPR1A
Gene Accession No. P37288
Gene Alias antidiuretic hormone receptor 1A; arginine vasopressin receptor 1A; arginine vasopressin receptor V1a; AVPR; AVPR V1a; AVPR1; Avpr1a; SCCL vasopressin subtype 1a receptor; V1a; V1a arginine vasopressin receptor; V1a receptor; V1a vasopressin receptor; V1aR; V1-vascular vasopressin receptor AVPR1A; Vascular/hepatic-type arginine vasopressin receptor; vasopressin V1a receptor; Vasopressin V1aR
Gene Symbols AVPR1A
Host Species Rabbit
Immunogen Peptide corresponding to Human AVPR1A (aa 398-418).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.