Learn More
AVANTOR MASTERFLEX (VWR LLC) AW-Lake Stainless steel meter-mounted digital flowmeter with integral display, 2.0 GPM
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3292836
Encompass
Precisely measured gear teeth deliver consistent ±0.5% accuracy even when measuring high-viscosity fluids¿ideal for measuring the flow of oils, grease, fuels, solvents, and other nonabrasive lubricating liquids. These positive displacement flowmeters are solidly built to provide excellent dynamic response for use in high-pressure applications. Gears and bearings withstand bidirectional flow without damage. Two 6-mm mounting holes let you secure the meter on a base plate, panel, or manifold. Available with a high-strength aluminum or 303 SS body. Pulse Output Flowmeters provide a NPN sourcing square wave pulse that is proportional to the flow rate. The peak-to-peak voltage of this square wave is the supply voltage provided minus two volts. &
Catalog No. NC1545692
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.