Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Awl-II-38.3 | 1135205-94-5 | 99.7% | 469.42 g·mol⁻¹ | C23H18F3N5O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1883210MG

Encompass_Preferred

AWL-II-38.3 is a small-molecule inhibitor of ephrin-A receptor A3 (EphA3) developed for research use. It exhibits potent kinase inhibitory activity against EphA3 with limited cellular activity against Src-family kinases and B-RAF, making it useful for target validation and biochemical assay development.

  • Potent EphA3 inhibition in biochemical assays.
  • Minimal off-target cellular activity against Src-family kinases and B-RAF.
  • High purity suitable for research applications.
  • Molecular weight 469.42 g·mol⁻¹ and formula C23H18F3N5O3.
  • Soluble in DMSO at high concentration for in vitro use.
  • Stable as a powder when stored at cold temperatures; follow solvent storage guidelines.
  • Solid, white to light yellow appearance for easy visual confirmation.
  • Provided in quantities appropriate for assay development and preclinical studies.

Catalog No. 50-002-98488


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.