Learn More
Medchemexpress LLC Axitinib-13C,d3 | 1261432-00-1 | 97.8% | 390.48 g/mol | C21 13C3 H15 D3 N4 O S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10065S5MG
Encompass_Preferred
Axitinib-13C,d3 is a stable isotope-labeled form of the tyrosine kinase inhibitor axitinib prepared for analytical and research applications. It contains 13C and deuterium labels that facilitate traceable quantitation and identification in mass spectrometry and related assays. Reported purity and molecular weight support its use as an analytical standard in method development and validation.
  • Isotopically labeled for analytical tracing and quantitation
  • Suitable as an internal standard in mass spectrometry workflows
  • High reported purity, appropriate for analytical applications
  • Available in small-mass packaging for method development
  • For research use only; not for human or veterinary use
Catalog No. 50-002-13260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.