Learn More
Invitrogen™ Axl Monoclonal Antibody (DS7HAXL), Brilliant Violet™ 421, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | Axl |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DS7HAXL |
| Concentration | 5 μL/Test |
| Conjugate | Brilliant Violet 421 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | AXL |
| Gene Accession No. | P30530 |
| Gene Alias | Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.