Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 0.6mL Conical Screw Cap Tubes: Presterilized

Catalog No. 14222647 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-647 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-647 Supplier Axygen™ Supplier No. SCT45CS
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY

Specifications

Cap Style Screw Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Closure Material Polypropylene
Capacity (Metric) 0.6 mL
Color Clear
Material Polypropylene
Format Non-looped
Includes O-ring
No. per Pack 500/Pk.
Packaging 500 Elangated Tubes and caps/Pk.; 8 Packs/Cs.
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.