Learn More
Description
For collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
- Available as presterilized and nonsterile microtubes
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached, with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Color: Clear
Specifications
Specifications
| Closure Color | Clear |
| Closure Material | Polypropylene |
| Capacity (Metric) | 0.6 mL |
| Color | Clear |
| Material | Polypropylene |
| Format | Non-looped |
| Includes | O-ring |
| Packaging | Bag |
| Sterility | Sterile |
| Closure Type | Screw Cap |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.