Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 096 Round, Deep-Well Storage Microplates, Clear, 2.0mL

Catalog No. 10999100 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
10-999-100 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-999-100 Supplier Axygen™ Supplier No. PDW20CCOVANCE

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Excellent chemical and low temperature resistance

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
  • 2mL deep-well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS 1-2004
Array 8 x 12
Color Clear
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Total Volume (Metric) Well 2 mL
Well Shape Round
No. per Pack 5
Temperature Range (Metric) Down to -80°C
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.