Learn More
Axygen™ 1.5 mL Conical Screw Cap Tubes
Catalog No. 14222565 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Assorted
Blue
Clear
Green
Orange
Red
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Closure Color
14-222-565 Sterile Assorted
14-222-569 Sterile Blue
14-222-570 Non-sterile Clear
14-222-571 Sterile Clear
14-222-576 Sterile Orange
14-222-574 Sterile Green
14-222-579 Sterile Red
14-222-606 Sterile Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-222-565 Supplier Axygen™ Supplier No. SCT150AS
Add to Cart
Add to Cart
With chemical-resistant O-ring
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Assorted
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Packaging Bag
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov