Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 12-Strip Flat PCR Tube Caps
GREENER_CHOICE

Catalog No. 14222300 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-300 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-300 Supplier Axygen™ Supplier No. PCR12FCPC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Axygen PCR tube strips (see PCR-0212-C). Manufactured from 99.9% pure virgin polypropylene.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Color Clear
Volume (Metric) 0.2 mL
Quantity 80 Pack
Closure Material Polypropylene
For Use With (Equipment) 12-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Strip
Product Type 12-Strip Flat PCR Tube Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.