Learn More
Axygen™ 120μm CyclerSeal Sealing Film for Storage and PCR Applications, Nonsterile
GREENER_CHOICE
Catalog No. 14222348 Shop All Axygen Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
14-222-348 Polypropylene
1 options
Catalog No. 14-222-348 Supplier Axygen™ Supplier No. PCRTS900
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
Axygen™ microplate sealing films are designed for applications ranging from PCR and Real-time PCR to ELISA and cell culture. They can be used in a wide range of temperatures, making them suitable for nearly any application.
  • This polypropylene film is the easiest and most cost-effective method for sealing plates
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen compression mat, eliminates well to well contamination and cross-over in PCR application
  • PCR-TS-900 is suitable for water-bath and other PCR application
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Sterility Non-sterile
No. per Case 250
No. per Pack 50
Product Type Sealing Film
Thickness 120 μm
WARNING: Cancer - www.P65Warnings.ca.gov