Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 165μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format

Catalog No. 14222898 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
165 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-222-898 165 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-898 Supplier Axygen™ Supplier No. VTF165LRS

For use with Agilent/Velocity11 automation systems

  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 165 μL
Filtered Filtered
Sterility Sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Maxymum Recovery™, non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Material Polyethylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.