Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Maxymum Recovery™ 20 μL Ultra Micro Tips

Catalog No. 14222758 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
14-222-758 Non-sterile Rack
14-222-757 Non-sterile Bulk
14-222-759 Sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-222-758 Supplier Axygen™ Supplier No. T400LR
Add to Cart
Add to Cart

Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are RNase-/DNase-free and nonpyrogenic. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic andRNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 20 μL
Sterility Non-sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point, Low Retention
Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment) Eppendorf, Gilson Pipetman P2, Gilson Pipetman P10
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.