Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 4- Strip PCR Tubes and Caps for Corbett Rotor-Gene™
GSA_VAGREENER_CHOICE

Catalog No. 14222247 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-247 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-247 Supplier Axygen™ Supplier No. PCR0104C
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easy snap and lock caps

Axygen™ 0.1mL PCR tubes are specifically designed for use with the Corbett Rotorgene system. There are 4 tubes per strip and 4 caps per strip.

  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene

Order Info

Tubes and caps are in strips of four and are in separate bags for ease of handling and labeling.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase and non-pyrogenic free
Closure Color Clear
Color Clear
Volume (Metric) 0.1 mL
Quantity 250/Pk.
Closure Material Polypropylene
For Use With (Equipment) Corbett™ Rotor-Gene™ system
Shape Conical
Sterility Non-sterile
Material Polypropylene
Closure Type Snap and Lock Plug Seal Flat Cap
Format Strip
Product Type 4-Strip PCR Tube And Flat Cap
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.