Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 96-well PCR Microplates

Catalog No. 14222330 Shop All Axygen Products
Change view
Click to view available options
Color:
Blue
Clear
Red
Skirt Style:
Full
Half
Unskirted
Format:
Flat Top
Single Notch Corner
Standard
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style Format
14-222-330 Red Full Standard
14-222-321 Clear Unskirted Standard
14-222-322 Clear Unskirted Flat Top
14-222-326 Clear Full Standard
14-222-324 Blue Full Standard
14-222-334 Clear Half Standard
14-222-340 Clear Half Single Notch Corner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-222-330 Supplier Axygen™ Supplier No. PCR96FSR

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.

Axygen™ 96 well elevated skirt PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 96-well Semi-Skirted PCR Plate
  • Single Notch Coner
  • Application/Compatibility: Standard thermal cycling, ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode No Barcode
    Certifications/Compliance RNase-, DNase-free
    Format Standard
    Array 8 x 12
    Color Red
    For Use With (Equipment) Standard 96 well blocks
    Skirt Style Full
    Material Polypropylene
    Volume (Metric) Well 200 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.