Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Biomek™ FX/NX Aerosol Barrier Filtered Robotic Tips

Catalog No. 14222128 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
20 μL
25 μL
30 μL
50 μL
165 μL
180 μL
200 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Extended Length, Low Retention
Automation, Paper Bottom, Round
Automation, Round
Extended Length, Automation
Fine Point, Automation
Low Retention, Round
Paper Bottom, Automation, Round
Round, Automation
Wide Orifice, Automation
Format:
Rack, 384-well
Rack, 96
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style Format
14-222-128 50 μL Sterile Low Retention, Round Rack, 96
14-222-121 20 μL Sterile Round, Automation Rack, 96
14-222-122 30 μL Sterile Fine Point, Automation Rack, 384-well
14-222-126 30 μL Sterile Extended Length, Automation Rack, 384-well
14-222-130 50 μL Sterile Round, Automation Rack, 96
14-222-113 165 μL Sterile Wide Orifice, Automation Rack, 96
14-222-117 180 μL Sterile Round, Automation Rack, 96
14-222-116 180 μL Non-sterile Paper Bottom, Automation, Round Rack, 96
14-222-118 200 μL Sterile Round, Automation Rack, 96
14-222-120 20 μL Sterile Low Retention, Round Rack, 96
14-222-123 30 μL Sterile Automation, Round Rack, 384-well
14-224-036 25 μL Sterile Automation, Round Rack, 384-well
14-222-125 30 μL Sterile Automation, Extended Length, Low Retention Rack, 384-well
14-222-129 50 μL Non-sterile Automation, Paper Bottom, Round Rack, 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 14-222-128 Supplier Axygen™ Supplier No. FXF50LRS

Clear polypropylene filter tips; autoclavable.

  • Tip volume range: 5 to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • non-pyrogenic
  • Available options:
    • With or without aerosol-resistant filters
    • non-sterile or sterile
    • Standard or Axygen™ Maxymum Recovery™ surface
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color Clear
Rack Color Green
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 96
Tip Style Low Retention, Round
Packaging 10 Racks/Pk, 5 Pks/Cs.
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Product Line Beckman Biomek
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.