Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Capless Tube for ABI Prism 310 Sequencer
GREENER_CHOICE

Catalog No. 14222861 Shop All Axygen Products
Change view
Click to view available options
Capacity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
14-222-861 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-222-861 Supplier Axygen™ Supplier No. TUB310NC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Capless tube for ABI Prism 310 sequencer

Axygen™ 0.5 mL polypropylene thin wall PCR tubes are designed to fit standard 96 well blocks.

  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Compatible with ABI-310 Instrument
  • Caps for these tubes are AM-SEPTA-310 Axygen™ AxyMatsTM Single Septa Cap Compatible with ABI-310 Sequencer, Nonsterile
  • TRUSTED_SUSTAINABILITY

    Specifications

    Capacity 0.5 mL
    For Use With (Equipment) ABI Prism 310 sequencer
    Product Type Capillary Sequencing Tube
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.