Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Caps for PCR Tubes
GREENER_CHOICE

Catalog No. 14223222 Shop All Axygen Products
Change view
Click to view available options
Color:
Assorted
Clear
MaxyClear
MaxyClear, w/writing surface
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type Product Type
14-223-222 Clear Domed Cap PCR Caps
14-222-265 Clear Domed Cap 8-Strip Domed PCR Tube Cap
14-222-263 Assorted Domed Cap 8-Strip Domed PCR Tube Cap
14-222-281 MaxyClear, w/writing surface Flat Cap 8-Strip Flat PCR Tube Cap with Writing Surface
14-222-302 MaxyClear Flat Cap 8-Strip Flat PCR Tube Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-223-222 Supplier Axygen™ Supplier No. PCR02CPC100
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

8-Strip domed or flat strip caps for Axygen PCR tubes

Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified RNase-DNase-free
Color Clear
Volume (Metric) 0.2 mL
For Use With (Equipment) 8 strip PCR tubes and 96-well microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format 1 x 8 Strip Caps
Product Type PCR Caps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.