Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Conical-Bottom Screw-Cap Tubes, Attached Clear Looped Caps

Catalog No. 14223071 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-223-071 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-223-071 Supplier Axygen™ Supplier No. SCT150LPC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Graduated tubes with conical bottom

  • Writing area for sample identification
  • Caps close with one hand

TRUSTED_SUSTAINABILITY

Specifications

Cap Style Screw Cap
Autoclavable Not Autoclavable
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Attached clear looped cap
Sterility Non-sterile
Bottom Shape Conical
Max. RCF 20,000 x g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.