Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Conical Centrifuge Tubes
GREENER_CHOICE

Catalog No. 14222961 Shop All Axygen Products
Change view
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Packaging
14-222-961 15 mL 25 Tubes and Caps/Rack, 20 Racks/Cs.
14-222-964 50 mL Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-222-961 Supplier Axygen™ Supplier No. SCT15ML500
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Graduated tubes with conical bottom

  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Cap Style Screw Cap
Autoclavable Not Autoclavable
Capacity (Metric) 15 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
Packaging 25 Tubes and Caps/Rack, 20 Racks/Cs.
Sterility Non-sterile
Bottom Shape Conical
Max. RCF 12,000 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.