Learn More
Axygen™ EasyPress Sealing Mat and Film Applicator
GREENER_CHOICE
Catalog No. 14222934 Shop All Axygen Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
14-222-934 1 Ea.
1 options
Catalog No. 14-222-934 Supplier Axygen™ Supplier No. ITEPR
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Provides simple and effective application of Axymats and ImpermaMats
Axygen™ EASY PRESS is a manual device which provides simple and effective application of Axymats and/or Impermamats to 24, 48, 96 or 384 well microplates, deep well plates, microtiter tube systems and PCR plates.The EASY PRESS can be quickly adjusted to handle any size plate of varying heights (384 PCR to 2mL deep well blocks) using the large thumbwheel.
  • Suitable for deep well plates, PCR microplates in 96 and 384 formats
  • Flexible-can be used with plates of various heights and types
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) Axymats, deep well plates, microplates, pcr plates
    No. per Pack 1 Ea.
    Product Type Sealer
    Quantity 1 Ea.
    WARNING: Cancer - www.P65Warnings.ca.gov