Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Low Profile 8-Strip PCR Tubes
GREENER_CHOICE

Catalog No. p-4495123 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
Product Type:
Thin Wall 8-Strip PCR Tube
Thin Wall 8-Strip PCR Tube with Real Time Strip Caps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
14-223-504 Clear Thin Wall 8-Strip PCR Tube
14-223-505 Clear Thin Wall 8-Strip PCR Tube with Real Time Strip Caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-223-504 Supplier Axygen™ Supplier No. PCR0108LPC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Axygen™ brand low profile 8-strip PCR tubes are ideal for low sample volume reactions.

Axygen™ Low Profile 8-Strip PCR tubes are ideal for low sample volume reactions. Using highly polished and precise molds, our tubes ensure precise thermal transfer and a leak-proof seal with a choice of strip caps. The low profile design allows reduced reaction time and evaporation due to less dead air volume. Tubes are available in white and clear with or without caps. Tubes are DNase-/RNase-free and nonpyrogenic.

 

  • Low profile design results in faster and more efficient PCR reactions
  • Ideal for 0.1 mL thermal cyclers
  • Available in white or clear
  • DNase-/RNase-free and Nonpyrogenic
  • Recommended volume range = 5 μL to 125 μL
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase and non-pyrogenic free
Color Clear
Volume (Metric) 0.1 mL
Quantity 125/Pk.
For Use With (Equipment) 0.1 mL thermal cycler for low sample volume reactions
Shape Conical
Sterility Non-sterile
Material Polypropylene
Format Strip
Product Type Thin Wall 8-Strip PCR Tube
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.